missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) sulfide, red

Product Code. 11346858
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11346858 500 g 500g
11336858 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346858 Supplier Thermo Scientific Alfa Aesar Supplier No. 013482.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 1344-48-5 | HgS | 232.65 g/mol

Mercury(II) sulfide, red is used chiefly as a pigment in paints, rubber and plastics. It is also used as a semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-48-5
Molecular Formula HgS
Molecular Weight (g/mol) 232.65
MDL Number MFCD00011046
InChI Key LGZXYFMMLRYXLK-UHFFFAOYSA-N
Synonym mercuric sulfide, mercury ii sulfide, vermilion, mercuric sulfide red, almaden, cinnabar, ethiops mineral, mercury sulfide, red cinnabar, mercury sulphide
PubChem CID 62402
SMILES [S--].[Hg++]

Specifications

Melting Point 583.5° (sublimation)
Color Red
Physical Form Powder
Quantity 500 g
Solubility Information Insoluble in water.
IUPAC Name mercury(2+) sulfanediide
Formula Weight 232.65
Odor Odorless
Appearance Red powder
Density 8.17
Chemical Name or Material Mercury(II) sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.