Learn More
Mercury(II) Sulfate, Extra Pure, SLR, Fisher Chemical
Brand: Fisher Chemical M/3200/50
33.20 GBP valid until 2019-12-31
Use promo code "13905" to get your promotional price.
Code : 21
Additional Details : CAS Number : 7783-35-9 Weight : 0.25000kg Transport : UN number : 1645 Chem class : 6.1 Pack group : II
Quantity | 250g |
---|
Chemical Identifiers
7783-35-9 | |
296.648 | |
DOBUSJIVSSJEDA-UHFFFAOYSA-L | |
24544 | |
[O-]S(=O)(=O)[O-].[Hg+2] |
HgO4S | |
11047 | |
mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate | |
mercury(2+);sulfate |
Specifications
7783-35-9 | |
11047 | |
DOBUSJIVSSJEDA-UHFFFAOYSA-L | |
mercury(2+);sulfate | |
24544 | |
Powder Solid | |
White | |
250g |
HgO4S | |
mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate | |
[O-]S(=O)(=O)[O-].[Hg+2] | |
296.648 | |
296.64g/mol | |
Plastic powder jar | |
1 |
Safety and Handling
- Mercury(II) sulfate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H310-Fatal in contact with skin.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H330-Fatal if inhaled.
- H373-May cause damage to organs.
- H300-Fatal if swallowed.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.