missing translation for 'onlineSavingsMsg'
Learn More

Mercury(Ii) Sulfate, ACS Reagent, ≥98.0%, Honeywell Fluka™

Product Code. 15671720 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15671720 500 g Poly Bottle 500g
15661720 100 g Poly Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15671720 Supplier Honeywell-Fluka Supplier No. 200093500G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS Reagent,≥98.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-35-9
Molecular Formula HgO4S
Molecular Weight (g/mol) 296.65
MDL Number MFCD00011047
InChI Key DOBUSJIVSSJEDA-UHFFFAOYSA-L
Synonym mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
PubChem CID 24544
IUPAC Name mercury(2+);sulfate
SMILES [Hg++].[O-]S([O-])(=O)=O

Specifications

Color White
Quantity 500 g
Linear Formula HgSO4
UN Number UN1645
Formula Weight 296.65g/mol
Percent Purity ≥98.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder/Crystals/Chunks
Density 7.56g/cu.cm
Chemical Name or Material Mercury(Ii) Sulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.