missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) sulfate, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog Number 11343358
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
7783-35-9
HgO4S
296.65
MFCD00011047
DOBUSJIVSSJEDA-UHFFFAOYSA-L
mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
24544
mercury(2+);sulfate
[Hg++].[O-]S([O-])(=O)=O
This item is not returnable. View return policy
7783-35-9
HgO4S
296.65
MFCD00011047
DOBUSJIVSSJEDA-UHFFFAOYSA-L
mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
24544
mercury(2+);sulfate
[Hg++].[O-]S([O-])(=O)=O

For Use As a Primary Battery Electrolyte, Catalyst in Conversion of Acetylene To Acetaldehyde

Mercury(II) sulfate is used for gold and silver extraction, and to make other mercury compounds. Used as catalyst in the conversion of acetylene to acetaldehyde and used as a electrolyte in primary battery. It is commonly used as a catalyst in oxymercuration-demercuration, a type of electrophilic addition reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point (decomposition)
Color White to Yellow
Physical Form Powder
Quantity 25 g
Assay Percent Range 98% min.
Solubility Information Soluble in hot sulfuric acid and sodium chloride solution. Insoluble in alcohol,acetone and ammonia.
Formula Weight 296.65
Grade ACS
Sensitivity Hygroscopic
Appearance White to pale yellow powder
Density 6.470 g/cm3 at 20°C
Chemical Name or Material Mercury(II) sulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'