missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) sulfate, ≥99.0% (precipitation titration), Purum p.a., Honeywell™

Product Code. 15677270 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Poly bottle
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15677270 250 g Poly bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15677270 Supplier Honeywell Research Chemicals Supplier No. 83372250G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Purum p.a.,≥99.0% (precipitation titration)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7783-35-9
Color White
pH Acidic
Physical Form Crystalline
Quantity 250 g
Molecular Formula HgO4S
Linear Formula HgSO4
MDL Number MFCD00011047
UN Number UN1645
Synonym mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
InChI Key DOBUSJIVSSJEDA-UHFFFAOYSA-L
SMILES [Hg++].[O-]S([O-])(=O)=O
IUPAC Name mercury(2+);sulfate
Molecular Weight (g/mol) 296.65
PubChem CID 24544
Formula Weight 296.65g/mol
Percent Purity ≥99.0% (precipitation titration)
Odor Odorless
Packaging Poly bottle
Density 7.56g/cu.cm at 20°C
Chemical Name or Material Mercury(II) sulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.