missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) selenide, 99.999% (metals basis)

Product Code. 11327567
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327567 5 g 5g
11317567 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327567 Supplier Thermo Scientific Alfa Aesar Supplier No. 012832.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20601-83-6 | HgSe | 279.56 g/mol

Mercury(II) selenide is used in semiconductors, infrared detectors, solar cells, transistors and amplifiers. It is also used as filters in some steel plants in order to remove mercury from exhaust gases. Further, it is used as an ohmic contact to wide-gap II-VI semiconductors such as zinc selenide or zinc oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20601-83-6
Molecular Formula HgSe
Molecular Weight (g/mol) 279.56
MDL Number MFCD00049743
InChI Key YQMLDSWXEQOSPP-UHFFFAOYSA-N
Synonym mercury selenide, mercury ii selenide, mercury-selenium complex, mercury monoselenide, mercury 2+ selenide, mercury ii selenide trace metals basis 1g
PubChem CID 88609
IUPAC Name selanylidenemercury
SMILES [Se]=[Hg]

Specifications

Melting Point (sublimation)
Color Gray
Physical Form Powder
Quantity 5 g
UN Number UN2025
Solubility Information Insoluble in water.
Formula Weight 279.55
Percent Purity 99.999%
Odor Odorless
Density 8.21 g/mL
Assay (metals basis)
Chemical Name or Material Mercury(II) selenide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.