missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) potassium iodide, Thermo Scientific Chemicals

Product Code. 11353718
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353718 50 g 50g
11373718 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11353718 Supplier Thermo Scientific Alfa Aesar Supplier No. 039307.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mercury(II) potassium iodide is used as a pharmaceutical intermediate. It is also used as reagents, chemical research and organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-33-7
Molecular Formula H4HgI4K2
Molecular Weight (g/mol) 790.44
MDL Number MFCD00049656
InChI Key ZCPVFAXYZYZMBC-UHFFFAOYSA-N
Synonym nessler's reagent, nessler reagent, mayer's reagent, channing's solution, caswell no. 695, potassium iodomercurate, mercury ii potassium iodide, mercury potassium iodide, potassium mercuric iodide, mercuric potassium iodide
PubChem CID 24542
ChEBI CHEBI:51568
IUPAC Name dipotassium;tetraiodomercury(2-)
SMILES [K+].[K+].I.I.I.I.[Hg--]

Specifications

Color Yellow
Physical Form Crystalline Aggregates
Quantity 50 g
Linear Formula K2HgI4
UN Number UN2025
Merck Index 14,7688
Formula Weight 786.4
Sensitivity Hygroscopic
Density 4.3g/mL
Chemical Name or Material Mercury(II) potassium iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.