missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) perchlorate trihydrate, Reagent Grade, Thermo Scientific™

Product Code. 11331738
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331738 25 g 25g
11321738 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331738 Supplier Thermo Scientific Alfa Aesar Supplier No. 011634.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73491-34-6
Molecular Formula Cl2H6HgO11
Molecular Weight (g/mol) 453.529
MDL Number MFCD00149735
InChI Key DFJWPGPUWHLJIJ-UHFFFAOYSA-L
Synonym mercury ii perchlorate trihydrate, mercury perchlorate-water 1/2/3, perchloric acid,mercury 2+ salt, trihydrate 9ci
PubChem CID 53471339
IUPAC Name mercury(2+);diperchlorate;trihydrate
SMILES O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Hg+2]

Specifications

Color White
Physical Form Crystalline
Quantity 25 g
Linear Formula Hg(ClO4)2·3H2O
UN Number UN3087
Solubility Information Soluble in water. Insoluble in benzene and hexane.
Formula Weight 453.54 (399.49 Anhydrous)
Grade Reagent
Chemical Name or Material Mercury(II) perchlorate trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.