missing translation for 'onlineSavingsMsg'
Learn More

Mercury(Ii) Oxide Yellow, ≥99.0%, ACS Reagent, Solstice

Product Code. 15672080 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15672080 500 g Glass Bottle 500g
15682080 5 g Glass Bottle 5g
15662080 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15672080 Supplier Honeywell-Fluka Supplier No. 221082500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21908-53-2
Molecular Formula HgO
Molecular Weight (g/mol) 216.59
MDL Number MFCD00011045
InChI Key UKWHYYKOEPRTIC-UHFFFAOYSA-N
Synonym mercuric oxide, mercury ii oxide, mercury oxide, mercuric oxide, red, red mercuric oxide, mercuric oxide, yellow, santar, red precipitate, mercury monoxide
IUPAC Name oxomercury
SMILES O=[Hg]

Specifications

Quantity 500 g
Linear Formula HgO
UN Number UN1641
Formula Weight 216.59g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Glass Bottle
Chemical Name or Material Mercury(Ii) Oxide Yellow
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercury(II) oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Mercury monoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.