missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II)Oxide, TRC

Product Code. 30240934
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240934 100 g 100g
30240560 25 g 25g
30240511 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240934 Supplier Toronto Research Chemicals Supplier No. TRCM225465100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21908-53-2
Molecular Formula Hg O
Molecular Weight (g/mol) 216.589
Synonym Mercuric oxide
IUPAC Name oxomercury
SMILES O=[Hg]

Specifications

Quantity 100 g
InChI Formula InChI=1S/Hg.O
Formula Weight 217.966
Chemical Name or Material Oxomercury
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.