missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) iodide, 99+%

Product Code. 11493167
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11493167

Brand: Thermo Scientific Alfa Aesar A16130.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7774-29-0 | HgI2 | 454.401 g/mol

Mercury(II) iodide is is used for preparation of Nessler's reagent, used for detection of presence of ammonia. It is a semiconductor material, used in some x-ray and gamma ray detection and imaging devices operating at room temperatures. In veterinary medicine, it is used in blister ointments in exostoses, bursal enlargement, etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mercury(II) iodide is is used for preparation of Nessler′s reagent, used for detection of presence of ammonia. It is a semiconductor material, used in some x-ray and gamma ray detection and imaging devices operating at room temperatures. In veterinary medicine, it is used in blister ointments in exostoses, bursal enlargement, etc.

Solubility
Insoluble in water. Slightly soluble in alcohol, ether, acetone, chloroform, ethyl acetate, olive oil and castor oil.

Notes
Light sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7774-29-0
Molecular Formula HgI2
Molecular Weight (g/mol) 454.401
MDL Number MFCD00011044
InChI Key YFDLHELOZYVNJE-UHFFFAOYSA-L
Synonym mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatum
PubChem CID 24485
ChEBI CHEBI:49659
IUPAC Name diiodomercury
SMILES I[Hg]I

Specifications

Boiling Point 350°C (sublimation)
Melting Point 259°C
Quantity 500 g
UN Number UN1638
Merck Index 14,5879
Solubility Information Insoluble in water. Slightly soluble in alcohol,ether,acetone,chloroform,ethyl acetate,olive oil and castor oil.
Formula Weight 454.4
Percent Purity ≥99%
Odor Odorless
Sensitivity Light sensitive
Density 6.28
Chemical Name or Material Mercury(II) iodide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.