missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) fluoride, 95%, Thermo Scientific Chemicals

Product Code. 11317227
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317227 10 g 10g
11327227 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317227 Supplier Thermo Scientific Alfa Aesar Supplier No. 011533.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mercury(II) fluoride is a selective fluorination agent. Organic compounds, such as triphenylacetic acid, triphenyl ethylene, and triethyl phosphite can be selectively fluorinated in dimethylsulfoxide/HgF 2 solutions under UV-visible illumination. CF2I2 was synthesized by fluorination of tetraiodomethane with mercury(II) fluoride and purified.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-39-3
Molecular Formula F2Hg
Molecular Weight (g/mol) 238.589
MDL Number MFCD00016139
InChI Key FMSYTQMJOCCCQS-UHFFFAOYSA-L
Synonym mercuric fluoride, mercury ii fluoride, mercury difluoride, mercury fluoride hgf2, hgf2
PubChem CID 82209
IUPAC Name difluoromercury
SMILES F[Hg]F

Specifications

Melting Point 645°C (decomposition)
Color Yellow
Physical Form Crystalline
Quantity 10 g
Linear Formula HgF2
UN Number UN2025
Merck Index 14,5878
Solubility Information Decomposes in water
Formula Weight 238.61
Percent Purity 95%
Odor Odorless
Sensitivity Moisture sensitive
Density 8.95 g/mL
Chemical Name or Material Mercury(II) fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.