missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury(II) fluoride, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Mercury(II) fluoride is a selective fluorination agent. Organic compounds, such as triphenylacetic acid, triphenyl ethylene, and triethyl phosphite can be selectively fluorinated in dimethylsulfoxide/HgF 2 solutions under UV-visible illumination. CF2I2 was synthesized by fluorination of tetraiodomethane with mercury(II) fluoride and purified.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 645°C (decomposition) |
Color | Yellow |
Physical Form | Crystalline |
Quantity | 50 g |
Linear Formula | HgF2 |
UN Number | UN2025 |
Merck Index | 14,5878 |
Solubility Information | Decomposes in water |
Formula Weight | 238.61 |
Percent Purity | 95% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'