missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) Chloride, TRC

Product Code. 30240611
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240611 5 g 5g
30180348 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30240611 Supplier Toronto Research Chemicals Supplier No. TRCM2575805G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7487-94-7
Molecular Formula Cl2 Hg
Molecular Weight (g/mol) 271.496
Synonym Mercury chloromercurate(II) (HgCl2) (6CI), Bichloride of mercury, Calochlor, Corrosive sublimate, CRC, Dichloromercury, Mercuric bichloride, Mercury bichloride, Mercury chloride, Mercury dichloride, Mercury dichloride (HgCl2), Mercury perchloride, Mercury(2+) chloride, Mercury(II) chloride, NSC 353255, Sublimate, Sulem
IUPAC Name dichloromercury
SMILES Cl[Hg]Cl

Specifications

Quantity 5 g
InChI Formula InChI=1S/2ClH.Hg/h2*1H;/q;;+2/p-2
Formula Weight 271.9083
Chemical Name or Material Mercuric chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.