missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) Chloride, 99%, Reagent Grade, Honeywell™

Product Code. p-7120556 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Poly bottle
Unit Size:
100g
1kg
25g
500g
Product Code. Quantity Packaging unitSize
15637930 25 g Poly bottle 25g
15617930 100 g Poly bottle 100g
15647930 500 g Poly bottle 500g
15627930 1 kg Poly bottle 1kg
4 options
This item is not returnable. View return policy

Product Code. 15637930

Brand: Honeywell Research Chemicals M113625G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7487-94-7
Molecular Formula Cl2Hg
Molecular Weight (g/mol) 271.49
MDL Number MFCD00011041
InChI Key LWJROJCJINYWOX-UHFFFAOYSA-L
Synonym mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2
PubChem CID 24085
ChEBI CHEBI:31823
IUPAC Name dichloromercury
SMILES [Cl-].[Cl-].[Hg++]

Specifications

Boiling Point 302°C
Melting Point 276°C
Color White
Physical Form Solid
Quantity 25 g
Linear Formula HgCl2
UN Number UN1624
Formula Weight 271.5g/mol
Percent Purity 99%
Odor Odorless
Packaging Poly bottle
Vapor Pressure 1.3mmHg (236°C)
Density 5.44g/cu.cm at 25°C
Chemical Name or Material Mercury(II) chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.