missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) bromide, ACS

Product Code. 11460111
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460111 100 g 100g
11450111 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460111 Supplier Thermo Scientific Alfa Aesar Supplier No. 044816.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-47-1 | Br2Hg | 360.4 g/mol

Mercury(II) bromide is used as a reagent in the Koenigs-Knorr reaction, which forms glycoside linkages on carbohydrates. It is also used to test for the presence of arsenic, as recommended by the Pharmacopoeia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-47-1
Molecular Formula Br2Hg
Molecular Weight (g/mol) 360.4
MDL Number MFCD00011040
InChI Key NGYIMTKLQULBOO-UHFFFAOYSA-L
Synonym mercuric bromide, mercury dibromide, mercury ii bromide, hgbr2, mercury bromide hgbr2, mercuric dibromide, mercury ii bromide 1:2, hsdb 829, hg2, mercury 2 bromide
PubChem CID 24612
ChEBI CHEBI:49639
IUPAC Name dibromomercury
SMILES Br[Hg]Br

Specifications

Boiling Point 322°C to 325°C (sublimation)
Melting Point 236°C
Color White
Physical Form Powder
Quantity 100 g
Solubility Information Soluble in hot alcohol,methanol,HCl,HBr
Formula Weight 360.4
Grade ACS
Odor Odorless
Sensitivity Light sensitive
Vapor Pressure 1mm Hg at 137°C
Density 6.05
Chemical Name or Material Mercury(II) bromide, For ACS analysis
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.