missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury(II) Acetate, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Mercury(II) Acetate |
| CAS | 1600-27-7 |
| Molecular Formula | C4 H6 Hg O4 |
| Quantity | 10 g |
| Synonym | Bis(acetyloxy)mercury, Diacetoxymercury, Mercuric Acetate, Mercuric Diacetate, Mercuric(II) Acetate, Mercury Acetate, Mercury Acetate (Hg(O2C2H3)2), Mercury Diacetate, Mercury(2+) Acetate, Mercury(II) Acetate, Mercury(II) Diacetate |
| InChI Formula | InChI=1S/2C2H4O2.Hg/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2 |
| SMILES | CC(=O)O[Hg]OC(=O)C |
| IUPAC Name | diacetyloxymercury |
| Molecular Weight (g/mol) | 318.68 |
| Formula Weight | 319.9972 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction