missing translation for 'onlineSavingsMsg'
Learn More

Mercury(I) nitrate dihydrate, 98%, for analysis

Product Code. 10424415 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Product Code. 10424415

Brand: Thermo Scientific Acros 213130010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

CAS: 7782-86-7 | Hg2N2O6·2H2O | 561.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-86-7
Molecular Formula H4Hg2N2O8
Molecular Weight (g/mol) 561.22
MDL Number MFCD00150812
InChI Key LSABZDVKJBWCBE-UHFFFAOYSA-N
IUPAC Name dimercury(1+) dihydrate dinitrate
SMILES O.O.[Hg+].[Hg+].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 70.0°C
Color White to Yellow
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 97.5% min
Linear Formula Hg2(NO3)2·2H2O
Fieser 06,360
Merck Index 13, 5923
Solubility Information Solubility in water: 20g/L (20°C).
Formula Weight 561.22
Percent Purity 98%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Mercury(I) nitrate dihydrate, p.a.
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercury(I) nitrate dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.