missing translation for 'onlineSavingsMsg'
Learn More

Mercury(I) chloride, 99.5%, Thermo Scientific Chemicals

Catalog Number 11334959
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
10112-91-1
Cl2Hg2
472.084
MFCD00011043
ZOMNIUBKTOKEHS-UHFFFAOYSA-L
mercurous chloride, calomel, mercury i chloride, mercury chloride hg2cl2, mercury subchloride, dimercury dichloride, calogreen, calotab, chlorure mercureux, ccris 3803
24956
CHEBI:33050
chloromercury
Cl[Hg].Cl[Hg]
This item is not returnable. View return policy
10112-91-1
Cl2Hg2
472.084
MFCD00011043
ZOMNIUBKTOKEHS-UHFFFAOYSA-L
mercurous chloride, calomel, mercury i chloride, mercury chloride hg2cl2, mercury subchloride, dimercury dichloride, calogreen, calotab, chlorure mercureux, ccris 3803
24956
CHEBI:33050
chloromercury
Cl[Hg].Cl[Hg]

For calomel electrodes, in ceramic painting, porcelain painting with gold

Mercury(I) chloride is used as calomel electrodes, in ceramic painting and porcelain painting with gold.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 400°C (sublimation)
Color White
Physical Form Crystalline
Quantity 100 g
UN Number UN2025
Merck Index 14,5894
Solubility Information Insoluble in water, alcohol, ether, dilute acids
Formula Weight 472.09
Percent Purity 99.5%
Odor Odorless
Sensitivity Light and moisture sensitive
Density 7.15 g/mL
Chemical Name or Material Mercury (I) chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'