missing translation for 'onlineSavingsMsg'
Learn More

Mercury Free Color Coded Buffer Solution pH 10 at 25 °C, Reagecon
BIOPHARMA

Product Code. 16470434 Shop All Reagecon Products
Click to view available options
Quantity:
1 L
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 16470434

Brand: Reagecon 110025CMF

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Guaranteed stability throughout entire shelf life even after opening the bottle, NIST traceable

  • Tested by ISO 17025 accredited test method

Specifications

Chemical Name or Material Color coded buffer solution
Color Blue
pH 10 ±0.01
Physical Form Liquid
Quantity 1 L
Packaging HDPE bottle GL45
Traceability to NIST Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.