missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercaptopurine Monohydrate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Mercaptopurine Monohydrate
Specifications
Specifications
| Chemical Name or Material | 6-Mercaptopurine monohydrate |
| CAS | 6112-76-1 |
| Analyte or Component Names | 6-Mercaptopurine monohydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C5 H4 N4 S . H2 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | 6H-Purine-6-thione, 1,2,3,9-tetrahydro-, hydrate (1:1), 6H-Purine-6-thione, 1,7-dihydro-, monohydrate (9CI), Purine-6-thiol, monohydrate (8CI), 6-Mercaptopurine monohydrate, 9H-Purine-6-thiol hydrate, Mercaptopurine hydrate, 7H-Purine-6-thiol hydrate, Azathioprine Impurity B as Monohydrate |
| InChI Formula | InChI=1S/C5H4N4S.H2O/c10-5-3-4(7-1-6-3)8-2-9-5;/h1-2H,(H2,6,7,8,9,10);1H2 |
| SMILES | O.Sc1ncnc2nc[nH]c12 |
| IUPAC Name | 7H-purine-6-thiol;hydrate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction