missing translation for 'onlineSavingsMsg'
Learn More

β-Mercaptoethylamine Hydrochloride, Fisher BioReagents

Product Code. 10586764
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10586764 100 g Amber Glass 100g
10174904 5 g Amber Glass 5g
10666765 25 g Amber Glass 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10586764
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
β-Mercaptoethylamine HCl is used as an antioxidant in several biochemical procedures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156-57-0
Molecular Formula C2H8ClNS
Molecular Weight (g/mol) 113.60
MDL Number MFCD00012904
InChI Key OGMADIBCHLQMIP-UHFFFAOYSA-N
Synonym cysteamine hydrochloride, 2-aminoethanethiol hydrochloride, cysteamine hcl, cysteaminium chloride, 2-mercaptoethylamine hydrochloride, 2-mercaptoethylamine hcl, bekaptan, mercaptamine hydrochloride, merkamin hydrochloride
PubChem CID 9082
IUPAC Name 2-aminoethanethiol;hydrochloride
SMILES [H+].[Cl-].NCCS

Specifications

Melting Point 66°C
Color White
pH 3.3 to 5.0
Assay Percent Range 98%
Packaging Amber Glass
Quantity 100 g
Merck Index 15, 2776
Formula Weight 113.61
Percent Purity ∼98%
Physical Form Solid
Chemical Name or Material β-Mercaptoethylamine Hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Aminoethanethiol hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.