missing translation for 'onlineSavingsMsg'
Learn More

Mercaptoacetic acid sodium salt, 97%

Product Code. 11323496
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11323496 25 g 25g
11333496 100 g 100g
11343496 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11323496 Supplier Thermo Scientific Alfa Aesar Supplier No. L14356.14
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 367-51-1 | C2H3NaO2S | 114.094 g/mol

Mercaptoacetic acid sodium salt is used as a reagent that protects tryptophan in amino acid analysis. It is a commonly used reagent for bacteriological research to maintain reducing conditions in media. It also protect enzymes against inactivation by maintaining protein thiol groups in the reduced state.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mercaptoacetic acid sodium salt is used as a reagent that protects tryptophan in amino acid analysis. It is a commonly used reagent for bacteriological research to maintain reducing conditions in media. It also protect enzymes against inactivation by maintaining protein thiol groups in the reduced state.

Solubility
Soluble in water (1000g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 20°C. Store under inert gas. It is hygroscopic in nature and sensitive to moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-51-1
Molecular Formula C2H3NaO2S
Molecular Weight (g/mol) 114.094
MDL Number MFCD00043386
InChI Key GNBVPFITFYNRCN-UHFFFAOYSA-M
Synonym sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate
PubChem CID 23690444
ChEBI CHEBI:86481
IUPAC Name sodium;2-sulfanylacetate
SMILES C(C(=O)[O-])S.[Na+]

Specifications

Melting Point >300°C
pH ∼6
Odor Stench
Quantity 25 g
UN Number UN3335
Beilstein 4569109
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,8692
Solubility Information Soluble in water (1000g/L at 20°C).
Formula Weight 114.1
Percent Purity 97%
Chemical Name or Material Mercaptoacetic acid sodium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.