missing translation for 'onlineSavingsMsg'
Learn More

Mercaptoacetic acid calcium salt trihydrate, 98%, Thermo Scientific™

Product Code. 15492357
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15492357 50 g 50g
15482357 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15492357 Supplier Thermo Scientific Alfa Aesar Supplier No. 045557.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65208-41-5
Molecular Formula C2H10CaO5S
Molecular Weight (g/mol) 186.24
MDL Number MFCD00150707
InChI Key GDECCIJZCWLFSB-UHFFFAOYSA-N
Synonym calcium,2-sulfidoacetate,trihydrate, thioglycolic acid calcium trihydrate
PubChem CID 131882808
IUPAC Name calcium;2-sulfanylacetic acid;trihydrate
SMILES O.O.O.[Ca].OC(=O)CS

Specifications

Melting Point 280°C to 290°C (decomposition)
Color White
Linear Formula (HSCH2CO2)Ca·3H2O
Quantity 50 g
Beilstein 3755791
Merck Index 14,1711
Solubility Information Soluble in water.
Formula Weight 276.34 (222.29 Anhydrous)
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Mercaptoacetic acid calcium salt trihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.