missing translation for 'onlineSavingsMsg'
Learn More

Mercaptoacetic Acid, 98%

Product Code. p-3736801 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
10mL
2.5 L
250mL
This item is not returnable. View return policy

Product Code. 10183410

Brand: Thermo Scientific Acros 125430100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68-11-1 | C2H4O2S | 92.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-11-1
Molecular Formula C2H4O2S
Molecular Weight (g/mol) 92.11
MDL Number MFCD00004876
InChI Key CWERGRDVMFNCDR-UHFFFAOYSA-N
Synonym mercaptoacetic acid, thioglycolic acid, 2-thioglycolic acid, acetic acid, mercapto, 2-mercaptoacetic acid, sulfanylacetic acid, thiovanic acid, mercaptoessigsaeure, thioglycollic acid, glycolic acid, thio
PubChem CID 1133
ChEBI CHEBI:30065
IUPAC Name 2-sulfanylacetic acid
SMILES C(C(=O)O)S

Specifications

Melting Point -16.0°C
Color Colorless
Density 1.3250g/mL
Boiling Point 220.0°C
Flash Point 126°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodimetry)
Packaging Glass bottle
Linear Formula HSCH2CO2H
Refractive Index 1.5030 to 1.5050
Quantity 10 mL
Fieser 01,1153; 07,366
Merck Index 15, 9489
Specific Gravity 1.325
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,ether,chloroform and benzene
Viscosity 6.55 Pas (20°C)
Formula Weight 92.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Mercaptoacetic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Mercaptoacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.