missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mer Mouse anti-Human, Clone: MERTK/3024, Novus Biologicals™
Mouse Monoclonal Antibody
£206.00 - £397.00
Specifications
| Antigen | Mer |
|---|---|
| Clone | MERTK/3024 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18398332
|
Bio-Techne
NBP3-13839-20UG |
20 μg |
£206.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18367445
|
Novus Biologicals
NBP3-13839-100UG |
100 μg |
£397.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mer Monoclonal antibody specifically detects Mer in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
| Mer | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cancer, Protein Kinase, Signal Transduction, Vision | |
| 10 mM PBS with 0.05% BSA | |
| c-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer | |
| Recombinant human Mer protein fragment (around aa 55-148) (exact sequence is proprietary) (Uniprot: Q12866) | |
| Primary | |
| Store at 4°C. |
| MERTK/3024 | |
| Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q12866 | |
| 10461 | |
| IgG2a | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title