missing translation for 'onlineSavingsMsg'
Learn More

Mer Antibody (MERTK/3024), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18393836 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393836 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393836 Supplier Novus Biologicals Supplier No. NBP314269AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mer Monoclonal antibody specifically detects Mer in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mer
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MERTK/3024
Conjugate Alexa Fluor 647
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias c-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer
Host Species Mouse
Immunogen Recombinant human Mer protein fragment (around aa 55-148) (exact sequence is proprietary) (Uniprot: Q12866)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Protein Kinase, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 10461
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.