missing translation for 'onlineSavingsMsg'
Learn More

Mequitazine, TRC

Product Code. 30134224
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134224 100 mg 100mg
30009676 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134224 Supplier Toronto Research Chemicals Supplier No. TRCM225110100MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29216-28-2
Molecular Formula C20 H22 N2 S
Molecular Weight (g/mol) 322.47
Synonym 10H-Phenothiazine, 10-(1-azabicyclo[2.2.2]oct-3-ylmethyl)-, Phenothiazine, 10-(3-quinuclidinylmethyl)- (8CI), Butix, LM 209, Mequitazine, Metaplexan, Mircol, NSC 303612, Nipolazin, Primalan, Quitadrill, Zesulan
IUPAC Name 10-(1-azabicyclo[2.2.2]octan-3-ylmethyl)phenothiazine
SMILES C(C1CN2CCC1CC2)N3c4ccccc4Sc5ccccc35

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H22N2S/c1-3-7-19-17(5-1)22(18-6-2-4-8-20(18)23-19)14-16-13-21-11-9-15(16)10-12-21/h1-8,15-16H,9-14H2
Formula Weight 322.1504
Chemical Name or Material Mequitazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.