missing translation for 'onlineSavingsMsg'
Learn More

Mequitazine, MedChemExpress

Product Code. 16494647
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494647 50 mg 50mg
16478856 100 mg 100mg
16471148 5 mg 5mg
16443816 25 mg 25mg
16408748 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494647 Supplier Medchem Express Supplier No. HYB2168
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Mequitazine is a potent, and long-acting histamine H1 antagonist.

Mequitazine is a potent, and long-acting histamine H1 antagonist.

Specifications

Chemical Name or Material Mequitazine
CAS 29216-28-2
Color White
Purity Grade Notes Research
Molecular Formula C20H22N2S
Quantity 50 mg
Synonym LM-209
Solubility Information DMSO : 16 mg/mL (49.62 mM; Need ultrasonic and warming)
SMILES C12=CC=CC=C1N(CC3CN4CCC3CC4)C5=C(C=CC=C5)S2
Molecular Weight (g/mol) 322.47
Formula Weight 322.47
Percent Purity 99.97%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.