missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mepyramine Maleate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Mepyramine maleate |
| CAS | 59-33-6 |
| Analyte or Component Names | Mepyramine maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H23 N3 O . C4 H4 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Mepyramine Maleate, N1-[(4-Methoxyphenyl)methyl]-N2,N2-dimethyl-N1-2-pyridinyl-1,2-ethanediamine (2Z)-2-butenedioate (1:1), N-[(4-Methoxyphenyl)methyl]-N',N'-dimethyl-N-2-pyridinyl-1,2-ethanediamine (2Z)-2-butenedioate (1:1), N-[(4-Methoxyphenyl)methyl]-N',N'-dimethyl-N-2-pyridinyl-1,2-ethanediamine (Z)-2-butenedioate (1:1), 2-[(2-Dimethylaminoethyl)(p-methoxybenzyl)amino]pyridine maleate, 2-[[2-(Dimethylamino)ethyl](p-methoxybenzyl)amino]pyridine Bimaleate, AH, Anisopyradamine, Antamine, Anthisan maleate, Antihist, Antisan, Diaminide maleate, Enrumay, Histalet Forte, Histalon, Histan, Histapyran, Histatex, Histavet P, Mepyramine hydrogen maleinate, Mepyramine maleate, Minihist, NSC 3604, Neo-Antergan maleate, Neoantergan maleate, Paramal, Paraminyl, Paraminyl maleate, Parmal, Prefrin A, Pymafed, Pyra Maleate, Pyramal, Pyranilamine maleate, Pyraninyl, Pyranisamine maleate, Pyrilamine maleate, Renstamin, Stamine, Stangen, Stangen maleate, Statomin maleate, Thylogen, Thylogen maleate, 1,2-Ethanediamine, N-[(4-methoxyphenyl)methyl]-N',N'-dimethyl-N-2-pyridinyl-, (2Z)-2-butenedioate (1:1) (9CI), 1,2-Ethanediamine, N-[(4-methoxyphenyl)methyl]-N',N'-dimethyl-N-2-pyridinyl-, (Z)-2-butenedioate (1:1), Pyridine, 2-[[2-(dimethylamino)ethyl](p-methoxybenzyl)amino]-, maleate (1:1) (8CI), 2-[(2-Dimethylaminoethyl)(p-methoxybenzyl)amino]-pyridine maleate, N-p-Methoxybenzyl-N',N'-dimethyl-N-alpha-pyridylethylenediamine maleate |
| InChI Formula | InChI=1S/C17H23N3O.C4H4O4/c1-19(2)12-13-20(17-6-4-5-11-18-17)14-15-7-9-16(21-3)10-8-15;5-3(6)1-2-4(7)8/h4-11H,12-14H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| Solution Type | Neat |
| SMILES | COc1ccc(CN(CCN(C)C)c2ccccn2)cc1.OC(=O)\C=C/C(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?