missing translation for 'onlineSavingsMsg'
Learn More

Meprin beta Subunit/MEP1B Antibody (289731) [DyLight 405], Novus Biologicals™

Product Code. 30736982 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736982 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736982 Supplier Novus Biologicals Supplier No. FAB28951E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Meprin beta Subunit/MEP1B Monoclonal antibody specifically detects Meprin beta Subunit/MEP1B in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Meprin beta Subunit/MEP1B
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 289731
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.24.63, endopeptidase-2, meprin A subunit beta, meprin A, beta, Meprin B, N-benzoyl-L-tyrosyl-p-amino-benzoic acid hydrolase beta subunit, N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit beta, PABA peptide hydrolase, PPH beta
Host Species Rat
Immunogen Human embryonic kidney cell line HEK293-derived recombinant human Meprin β Subunit/MEP1B, Thr23-Ser293, Accession # Q16820
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 4225
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.