missing translation for 'onlineSavingsMsg'
Learn More

Meprednisone, Mikromol™

Product Code. 18040650
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
18040650 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 18040650

Brand: Mikromol MM3686.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Meprednisone
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Meprednisone
CAS 1247-42-3
Analyte or Component Names Meprednisone
Shipping Condition Room Temperature
Molecular Formula C22 H28 O5
Certifications/Compliance ISO 17034
Synonym Beta, Betalone, Betanisona, Betapar, Betapred, Bitanisone, Deltacortene Beta, Deltisona B, Lepicortin-Beta, Meprednisone, Methylprednisone, NSC 527579, Sch 4358, Dexamethasone Imp. J (EP), Pregna-1,4-diene-3,11,20-trione, 17,21-dihydroxy-16-methyl-, (16beta)-, Pregna-1,4-diene-3,11,20-trione, 17,21-dihydroxy-16beta-methyl- (6CI,7CI,8CI), (16beta)-17,21-Dihydroxy-16-methylpregna-1,4-diene-3,11,20-trione, 16beta-Methylprednisone, 17,21-Dihydroxy-16beta-methylpregna-1,4-diene-3,11,20-trione
InChI Formula InChI=1S/C22H28O5/c1-12-8-16-15-5-4-13-9-14(24)6-7-20(13,2)19(15)17(25)10-21(16,3)22(12,27)18(26)11-23/h6-7,9,12,15-16,19,23,27H,4-5,8,10-11H2,1-3H3/t12-,15-,16-,19+,20-,21-,22-/m0/s1
SMILES C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3C(=O)C[C@]2(C)[C@@]1(O)C(=O)CO
IUPAC Name (8S,9S,10R,13S,14S,16S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,9,12,14,15,16-octahydrocyclopenta[a]phenanthrene-3,11-dione
Molecular Weight (g/mol) 372.45
Quantity 250 mg
Formula Weight 372.1937 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.