missing translation for 'onlineSavingsMsg'
Learn More

Mephenytoin, LoGiCal

Product Code. 30461644
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. Quantity unitSize
30461644 5 mg 5mg
1 options
This item is not returnable. View return policy

Product Code. 30461644

Brand: LoGiCal LGCFOR0348.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mephenytoin
CAS 50-12-4
Analyte or Component Names Mephenytoin
Shipping Condition Room Temperature
Molecular Formula C12 H14 N2 O2
Certifications/Compliance ISO 9001
Synonym Mephenytoin, 2,4-Imidazolidinedione, 5-ethyl-3-methyl-5-phenyl-Hydantoin, 5-ethyl-3-methyl-5-phenyl- (6CI,7CI,8CI), (±)-5-Ethyl-3-methyl-5-phenylhydantoin, (±)-Mephenytoin, (±)-Mesantoin, 3-Ethylnirvanol, 3-Methyl-5-ethyl-5-phenyl-2,4-imidazolidinedione, 3-Methyl-5-ethyl-5-phenylhydantoin, 5-Ethyl-3-methyl-5-phenyl-2,4(3H,5H)-imidazoledione, 5-Ethyl-3-methyl-5-phenylhydantoin, 5-Ethyl-3-methyl-5-phenylimidazolidin-2,4-dione, DL-Mephenytoin, Epiazin, Epilan, Fenantoin, Gerot-Epilan, Insulton, Mephenytoin, Mesantoin, Mesdontoin, Mesontoin, Methoin, Methylphenetoin, Metydan, NSC 34652, Phenantoin, Phenylethylmethylhydantoin, Sacerno, Sedantional, Sedantoin, Triantoin
InChI Formula InChI=1S/C12H14N2O2/c1-3-12(9-7-5-4-6-8-9)10(15)14(2)11(16)13-12/h4-8H,3H2,1-2H3,(H,13,16)
Solution Type Neat
SMILES CCC1(NC(=O)N(C)C1=O)c2ccccc2
IUPAC Name 5-ethyl-3-methyl-5-phenylimidazolidine-2,4-dione
Industry Type Clinical Testing
Molecular Weight (g/mol) 218.25
Quantity 5 mg
Formula Weight 218.1055 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.