missing translation for 'onlineSavingsMsg'
Learn More

Mephenytoin 1.0 mg/ml in Methanol, LoGiCal

Product Code. 30461847
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
30461847 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 30461847

Brand: LoGiCal LGCAMP0348.0001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Mephenytoin 1.0 mg/ml in Methanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mephenytoin
CAS 50-12-4
Analyte or Component Names Mephenytoin
Shipping Condition Ice Pack (-20°C)
Molecular Formula C12 H14 N2 O2
Certifications/Compliance ISO 17034
Synonym Mephenytoin, 2,4-Imidazolidinedione, 5-ethyl-3-methyl-5-phenyl-Hydantoin, 5-ethyl-3-methyl-5-phenyl- (6 CI,7 CI,8 CI), (±)-5-Ethyl-3-methyl-5-phenylhydantoin, (±)-Mephenytoin, (±)-Mesantoin, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione, 5-Ethyl-3-methyl-5-phenyl-2,4(3 H,5 H)-imidazoledione
InChI Formula InChI=1 S/C12H14N2O2/c1-3-12(9-7-5-4-6-8-9)10(15)14(2)11(16)13-12/h4-8 H,3H2,1-2H3,(H,13,16)
SMILES CCC1(NC(=O)N(C)C1=O)c2ccccc2
IUPAC Name 5-ethyl-3-methyl-5-phenylimidazolidine-2,4-dione
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 218.25
Quantity 1 mL
Formula Weight 218.1055 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.