missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEPCE Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | MEPCE |
|---|---|
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18305996
|
Novus Biologicals
NBP3-05056-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18382204
|
Novus Biologicals
NBP3-05056-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MEPCE Polyclonal specifically detects MEPCE in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| MEPCE | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 56257 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| BCDIN37SK snRNA methylphosphate capping enzyme, bicoid-interacting 3, homolog, Bin3 homolog, bin3, bicoid-interacting 3, homolog (Drosophila), EC 2.1.1.-, FLJ20257, methylphosphate capping enzyme | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 470-689 of human MEPCE (NP_062552.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title