missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEPCE Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562438
This item is not returnable.
View return policy
Description
Immunogen sequence: AQTPEYDVVL CLSLTKWVHL NWGDEGLKRM FRRIYRHLRP GGILVLEPQP WSSYGKRKTL TETIYKNYYR IQLKPEQFSS YLTSPDVGFS SYELVATPHN TSKGFQRPVY Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.
S-adenosyl-L-methionine-dependent methyltransferase that adds a methylphosphate cap at the 5'-end of 7SK snRNA (7SK RNA), leading to stabilize it (PubMed:17643375, PubMed:19906723, PubMed:30559425). Also has a non-enzymatic function as part of the 7SK RNP complex: the 7SK RNP complex sequesters the positive transcription elongation factor b (P-TEFb) in a large inactive 7SK RNP complex preventing RNA polymerase II phosphorylation and subsequent transcriptional elongation (PubMed:17643375). The 7SK RNP complex also promotes snRNA gene transcription by RNA polymerase II via interaction with the little elongation complex (LEC) (PubMed:28254838). In the 7SK RNP complex, MEPCE is required to stabilize 7SK RNA and facilitate the assembly of 7SK RNP complex (PubMed:19906723). MEPCE has a non-enzymatic function in the 7SK RNP complex; interaction with LARP7 within the 7SK RNP complex occluding its catalytic center (PubMed:19906723). [UniProt].
Specifications
| MEPCE | |
| Polyclonal | |
| Unconjugated | |
| mepce | |
| 7SK snRNA methylphosphate capping enzyme; BCDIN3; Bicoid-interacting protein 3 homolog; bin3 homolog; bin3, bicoid-interacting 3; bin3, bicoid-interacting 3, homolog; Bipl1; D5Wsu46e; fc59f06; FLJ20257; Mepce; methylphosphate capping enzyme; si:ch211-283h6.1; wu:fc59f06; zBCDIN3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56257 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7L2J0 | |
| mepce | |
| Recombinant protein corresponding to Human MEPCE. Recombinant protein control fragment (Product #RP-92757). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction