missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MEPCE Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29493
This item is not returnable.
View return policy
Description
MEPCE Polyclonal antibody specifically detects MEPCE in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation
Specifications
| MEPCE | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 56257 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| BCDIN37SK snRNA methylphosphate capping enzyme, bicoid-interacting 3, homolog, Bin3 homolog, bin3, bicoid-interacting 3, homolog (Drosophila), EC 2.1.1.-, FLJ20257, methylphosphate capping enzyme | |
| Between 639 and 689 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction