missing translation for 'onlineSavingsMsg'
Få mere at vide

MEPCE Antibody, Novus Biologicals™

Artikelnummer. 18405122 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
0.1 mL
25 μL
Pakningsstørrelse:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
18405122 25 μL 25µL
18149222 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18405122 Leverandør Novus Biologicals Leverandørnr. NBP23361925ul

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

MEPCE Polyclonal specifically detects MEPCE in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen MEPCE
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q7L2J0
Gene Alias BCDIN37SK snRNA methylphosphate capping enzyme, bicoid-interacting 3, homolog, Bin3 homolog, bin3, bicoid-interacting 3, homolog (Drosophila), EC 2.1.1.-, FLJ20257, methylphosphate capping enzyme
Gene Symbols MEPCE
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: AQTPEYDVVLCLSLTKWVHLNWGDEGLKRMFRRIYRHLRPGGILVLEPQPWSSYGKRKTLTETIYKNYYRIQLKPEQFSSYLTSPDVGFSSYELVATPHNTSKGFQRPVY
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56257
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Vis mere Vis mindre

For Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.