missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEP1A Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547562
This item is not returnable.
View return policy
Description
In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant human MEP1A is observed and 5% cross-reactivity with recombinant mouse MEP1B is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
MEP1A (Meprin A Subunit Alpha) is a Protein Coding gene. Among its related pathways are Collagen chain trimerization. Gene Ontology (GO) annotations related to this gene include metalloendopeptidase activity and metallopeptidase activity.
Specifications
| MEP1A | |
| Polyclonal | |
| Unconjugated | |
| MEP1A | |
| AI098089; AW107200; bA268F1.1 (meprin A alpha (PABA peptide hydrolase)); E-24.18; Endopeptidase-2; endopeptidase-24.18 subunit alpha; Mep1; Mep-1; MEP1A; Mep-1a; meprin 1 alpha; meprin A alpha; meprin A alpha-subunit; meprin A subunit alpha; meprin A, alpha (PABA peptide hydrolase); meprin alpha; N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha; PABA peptide hydrolase; PPH alpha; PPHA | |
| Goat | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17287 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P28825 | |
| MEP1A | |
| Sf 21-derived recombinant mouse Meprin alpha Subunit/MEP1A Val34-Arg615. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction