missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEOX 2 Antibody (OTI4F6), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72650DL594
This item is not returnable.
View return policy
Description
MEOX 2 Monoclonal antibody specifically detects MEOX 2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MEOX 2 | |
| Monoclonal | |
| DyLight 594 | |
| growth arrest-specific homeo box, Growth arrest-specific homeobox, homeobox protein MOX-2, mesenchyme homeo box 2 (growth arrest-specific homeo box), mesenchyme homeobox 2GAX, MOX2mesenchyme homeobox 2 (growth arrest-specific homeo box) | |
| Full length recombinant protein of human MEOX 2 (NP_005915) produced in E.coli. | |
| 0.1 mL | |
| Neuroscience | |
| 4223 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot | |
| OTI4F6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction