missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEOX 2 Antibody (OTI4F6) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72650
This item is not returnable.
View return policy
Description
MEOX 2 Monoclonal specifically detects MEOX 2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| MEOX 2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000 | |
| growth arrest-specific homeo box, Growth arrest-specific homeobox, homeobox protein MOX-2, mesenchyme homeo box 2 (growth arrest-specific homeo box), mesenchyme homeobox 2GAX, MOX2mesenchyme homeobox 2 (growth arrest-specific homeo box) | |
| Mouse | |
| 33.4 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| OTI4F6 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| MEOX2 | |
| Full length human recombinant protein of human MEOX2 (NP_005915) produced in E.coli. | |
| Affinity Purified | |
| Neuroscience | |
| 4223 | |
| Human, Mouse, Rat | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction