missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MEOX 2 Monoclonal antibody specifically detects MEOX 2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
Specifications
Specifications
| Antigen | MEOX 2 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Sandwich ELISA |
| Classification | Monoclonal |
| Clone | 4E10 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | AAH17021 |
| Gene Alias | growth arrest-specific homeo box, Growth arrest-specific homeobox, homeobox protein MOX-2, mesenchyme homeo box 2 (growth arrest-specific homeo box), mesenchyme homeobox 2GAX, MOX2mesenchyme homeobox 2 (growth arrest-specific homeo box) |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?