missing translation for 'onlineSavingsMsg'
Learn More

Menthone, mixture of isomers, 98%

Product Code. 11403248
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403248 50 g 50g
11413248 250 g 250g
11493238 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403248 Supplier Thermo Scientific Alfa Aesar Supplier No. A17665.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10458-14-7 | C10H18O | 154.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10458-14-7
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00062998
InChI Key NFLGAXVYCFJBMK-UHFFFAOYSA-N
Synonym isomenthone, p-menthan-3-one, 2-isopropyl-5-methylcyclohexanone, menthone, cyclohexanone, 5-methyl-2-1-methylethyl, 5-methyl-2-1-methylethyl cyclohexanone, p-menthanone, 2-isopropyl-5-methyl-cyclohexanone, p-menthan-3-one, dl, 5-methyl-2-isopropyl cyclohexanone
PubChem CID 6986
ChEBI CHEBI:36742
IUPAC Name 5-methyl-2-propan-2-ylcyclohexan-1-one
SMILES CC1CCC(C(=O)C1)C(C)C

Specifications

Density 0.894
Boiling Point ∼207°C
Flash Point 72°C (162°F)
Quantity 50 g
Formula Weight 154.25
Percent Purity 98%
Chemical Name or Material Menthone, mixture of isomers

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.