missing translation for 'onlineSavingsMsg'
Learn More

Menthol Racemic, Mikromol™

Product Code. 17980836
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17980836 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17980836 Supplier Mikromol Supplier No. MM3006.00
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Menthol Racemic
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Menthol Racemic
CAS 89-78-1
Analyte or Component Names Menthol Racemic
Shipping Condition Room Temperature
Molecular Formula C10 H20 O
Certifications/Compliance ISO 17034
Synonym Menthol racemic, (1RS,2SR,5RS)-5-methyl-2-(1-methylethyl)cyclohexanol, Cyclohexanol, 5-methyl-2-(1-methylethyl)-, (1R,2S,5R)-rel-, Cyclohexanol, 5-methyl-2-(1-methylethyl)-, (1α,2β,5α)-, Menthol, cis-1,3,trans-1,4- (8CI), Menthol, dl- (6CI), (1R,2S,5R)-rel-5-Methyl-2-(1-methylethyl)cyclohexanol, (±)-Menthol, DL-Menthol, Fisherman's Friend Lozenges, Hexahydrothymol, Meggezone, Menthacamphor, Menthol, Menthomenthol, NSC 2603, Peppermint camphor, Racementhol, Racemic menthol, Therapeutic Mineral Ice, Thymomenthol, dl-Menthol, rac-Menthol, rel-(1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexanol
InChI Formula InChI=1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3/t8-,9+,10-/m1/s1
SMILES CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O
IUPAC Name (1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol
Molecular Weight (g/mol) 156.27
Quantity 250 mg
Formula Weight 156.1514 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.