missing translation for 'onlineSavingsMsg'
Learn More

Menthofuran, 95%

Product Code. 10432815 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10432815 50 g Glass bottle 50g
10722022 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10432815 Supplier Thermo Scientific Acros Supplier No. 413560500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 494-90-6 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 494-90-6
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00041851
InChI Key YGWKXXYGDYYFJU-UHFFFAOYSA-N
Synonym menthofuran, menthofurane, 3,9-epoxy-p-mentha-3,8-diene, benzofuran, 4,5,6,7-tetrahydro-3,6-dimethyl, p-mentha-3,8-diene, 3,9-epoxy, 4,5,6,7-tetrahydro-3,6-dimethylbenzofuran, fema no. 3235, 4,5,6,7-tetrahydro-3,6-dimethylcoumarone, r-menthofuran, 3,6-dimethyl-4,5,6,7-tetrahydrobenzo 2,1-b furan
PubChem CID 329983
ChEBI CHEBI:50542
IUPAC Name 3,6-dimethyl-4,5,6,7-tetrahydro-1-benzofuran
SMILES CC1CCC2=C(C1)OC=C2C

Specifications

Color Yellow
Density 0.9700g/mL
Boiling Point 87.0°C (15.0 mmHg)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4832 to 1.4852
Quantity 50 g
Specific Gravity 0.97
Solubility Information Solubility in water: insoluble
Formula Weight 150.22
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Menthofuran
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Menthofuran
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.