missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Meningococcus Agglutinating Sera

Product Code. 13285589
Change view
Click to view available options
Organism Group:
Group X
Group Y
Group Z
Polyvalent Groups X-Z, W135
Unit Size:
2mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Organism Group Unit Size
13285589 Group Z 2mL
13235589 Polyvalent Groups X-Z, W135 2mL
13265589 Group X 2mL
13275589 Group Y 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 13285589 Supplier Thermo Scientific™ Supplier No. R30167301
  • ONLINE EXCLUSIVE PRICE
    £305.00
    £298.20 / 2mL
    Save £6.80
    2% Off
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Serologically identify Neisseria meningitidis by rapid agglutination test.

Serologically identify Neisseria meningitidis Groups A, C, D, X, Y, Z and W135 for epidemiological and diagnostic purposes with Thermo Scientific™ Remel™ Meningococcus Agglutinating Sera.

Use Meningococcus Agglutinating Sera for presumptive identification of Neisseria meningitidis grown on culture media.

  • Simple, easy-to-use tests
  • Identifies major groups of Neisseria meningitidis serotypes
  • Provides accurate, reliable and rapid results
  • Serological tests are based on the fact that antibodies in serum, produced in response to exposure to bacterial antigens, will agglutinate with bacteria carrying homologous antigens.

    Serological tests are intended for screening purposes and should augment, not replace, culture procedures.

    The monovalent meningococcus agglutinating sera are suitable for use in slide agglutination tests. An alternative method of typing, counter immunoelectroophoresis is also described in literature1,2,3,4. The polyvalent agglutinating sera are for slide use only.

    Not all products are available for sale in all territories. Please inquire.

    Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.

    TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage 2°C to 8°C
    Description Meningococcus Agglutinating Sera
    Detectable Analytes N. gonorrhoeae
    Sample Type Serum
    Type Agglutinating Sera
    Includes Meningococcus Agglutinating Sera 1 dropper bottle
    Organism Group Group Z
    Quantity 2 mL
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.