missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Menin Antibody (JE30-97), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32588
This item is not returnable.
View return policy
Description
Menin Monoclonal antibody specifically detects Menin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Menin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 | |
| menin, multiple, multiple endocrine neoplasia I | |
| Synthetic peptide within Human Menin aa 566-615/ 615. (Uniprot: O00255) | |
| 100 μL | |
| Cancer, DNA Repair, Transcription Factors and Regulators, Tumor Suppressors | |
| 4221 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| JE30-97 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction