missing translation for 'onlineSavingsMsg'
Learn More

Menaquinone-7, MedChemExpress

Product Code. 16484638
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484638 10 mg 10mg
16475476 100 mg 100mg
16457357 5 mg 5mg
16410937 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16484638 Supplier Medchem Express Supplier No. HY112499
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Menaquinone-7 (Vitamin K2-7), belongs to a class of K2-vitamin homologs, is originally discovered as the anti-hemorrhagic factors.

Menaquinone-7 (Vitamin K2-7) is identified as the most bioactive cofactor for the carboxylation reaction of Gla-proteins . Supplementation with Menaquinone-7 (Vitamin K2-7) is a pharmacological option for activating matrix Gla protein and intervening in the progression of calcific aortic valve stenosis (CAVS).

Specifications

Chemical Name or Material Menaquinone-7
CAS 2124-57-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C46H64O2
Quantity 10 mg
Synonym Vitamin K2-7 Vitamin K2(35) Vitamin MK-7
Solubility Information DMSO : 5 mg/mL (7.70 mM; Need ultrasonic and warming)
SMILES O=C1C(C/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)\C)=C(C)C(C2=C1C=CC=C2)=O
Molecular Weight (g/mol) 649.0
Formula Weight 649.0
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.