missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Menaquinone-7, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Menaquinone-7 (Vitamin K2-7) is identified as the most bioactive cofactor for the carboxylation reaction of Gla-proteins . Supplementation with Menaquinone-7 (Vitamin K2-7) is a pharmacological option for activating matrix Gla protein and intervening in the progression of calcific aortic valve stenosis (CAVS).
Specifications
Specifications
| Chemical Name or Material | Menaquinone-7 |
| CAS | 2124-57-4 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C46H64O2 |
| Quantity | 100 mg |
| Synonym | Vitamin K2-7 Vitamin K2(35) Vitamin MK-7 |
| Solubility Information | DMSO : 5 mg/mL (7.70 mM; Need ultrasonic and warming) |
| SMILES | O=C1C(C/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)\C)=C(C)C(C2=C1C=CC=C2)=O |
| Molecular Weight (g/mol) | 649.0 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction