missing translation for 'onlineSavingsMsg'
Learn More

Menaquinone 7-d7 (100μg/mL in Acetonitrile) 90%, TRC

Product Code. 30390023
Change view
Click to view available options
Quantity:
1x1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390023 1x1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390023 Supplier Toronto Research Chemicals Supplier No. TRCKIT61381X1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Menaquinone 7-d7
CAS 1233937-31-9
Molecular Formula C46H57D7O2
Quantity 1x1 mL
Synonym (all-E)-2-(3,7,11,15,19,23,27-Heptamethyl-2,6,10,14,18,22,26-octacosaheptaenyl)-3-methyl-1,4-naphthalenedione-d7, Vitamin K2(35)-d7, Menaquinone K7-d7, Vitamin MK 7-d7
InChI Formula InChI=1S/C46H64O2/c1-34(2)18-12-19-35(3)20-13-21-36(4)22-14-23-37(5)24-15-25-38(6)26-16-27-39(7)28-17-29-40(8)32-33-42-41(9)45(47)43-30-10-11-31-44(43)46(42)48/h10-11,18,20,22,24,26,28,30-32H,12-17,19,21,23,25,27,29,33H2,1-9H3/b35-20+,36-22+,37-24+,38-26+,39-28+,40-32+/i9D3,10D,11D,30D,31D
SMILES O=C1C(C/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/C)=C(C([2H])([2H])[2H])C(C2=C([2H])C([2H])=C([2H])C([2H])=C21)=O
Molecular Weight (g/mol) 656.04
Formula Weight 655.5346
Concentration or Composition (by Analyte or Components) 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.